Learn More
Selleck Chemical LLC Bosentan S4220-50mg
SDP
Catalog No. 503109118
Encompass_Preferred
Bosentan is an endothelin (ET) receptor antagonist for ET-A and ET-B with Ki of 4 7 nM and 95 nM respectively
Catalog No. 50-310-9118 Supplier Selleck Chemical LLC Supplier No. S422050MG
Bosentan is an endothelin (ET) receptor antagonist for ET-A and ET-B with Ki of 4 7 nM and 95 nM respectively