Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bosentan-d4 1mg | 1065472-77-6 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1154171MG

Encompass_Preferred

Bosentan-d4 is the deuterium-labeled (d4) analogue of bosentan, a competitive, dual antagonist of endothelin-1 at ETA and ETB receptors. It is used primarily as an analytical internal standard and tracer for LC-MS/MS, pharmacokinetic, and ADME studies, providing matched chromatographic behavior with a distinct mass.

  • Deuterium-labeled (d4) analogue for accurate mass spectrometry quantitation.
  • Used as an internal standard in pharmacokinetic and ADME studies.
  • Chemical formula C27H29N5O6S and molecular weight 555.6 g/mol.
  • Purity 98% suitable for analytical applications.
  • Acts as a dual ETA/ETB endothelin-1 receptor antagonist (analogue of bosentan).

Catalog No. 50-002-28454


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.