Learn More
Cayman Chemical Bosentan-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1065472-77-6, 5 mg
SDP
Catalog No. 502987040
Encompass_Preferred
An internal standard for the quantification of bosentan by GC- or LC-MS
Catalog No. 50-298-7040 Supplier Cayman Chemical Supplier No. 220975 MG
An internal standard for the quantification of bosentan by GC- or LC-MS