Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bosentan (hydrate) | 157212-55-0 | 99.9% | C27H31N5O7S | 1 ML
SDP

Catalog No. 5000432593
Encompass_Preferred

Bosentan hydrate is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors, with Ki values of 4.7 nM and 95 nM respectively, in human smooth muscle cells. This product is for research use only.

  • Competitive and dual antagonist of endothelin-1 (ET)
  • Targets ETA and ETB receptors
  • Ki values of 4.7 nM for ETA receptor and 95 nM for ETB receptor
  • For research use only
  • Appearance: Off-white to light yellow solid
  • Melting point: 114-118°C

Catalog No. 50-004-32593 Supplier Medchemexpress LLC Supplier No. HYA0013A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bosentan hydrate is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors, with Ki values of 4.7 nM and 95 nM respectively, in human smooth muscle cells. This product is for research use only.

  • Competitive and dual antagonist of endothelin-1 (ET)
  • Targets ETA and ETB receptors
  • Ki values of 4.7 nM for ETA receptor and 95 nM for ETB receptor
  • For research use only
  • Appearance: Off-white to light yellow solid
  • Melting point: 114-118°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.