Learn More
Apexbio Technology LLC Bosentan Hydrate, 500mg. Cas: 157212-55-0 MFCD: MFCD09751188. endothelin receptor antagonist
SDP
Catalog No. 501905250
Encompass_Preferred
IC50: Inhibiting endothelin receptor A and B with an IC50 of 15.1 1.6 M in P388/dx cells.A sulfonamide-derived, competitive and specific endothelin receptor antagonist with a relatively higher affinity to the endothelin A receptor than endothelin B receptor. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5250 Supplier Apexbio Technology LLC Supplier No. B1521500
IC50: Inhibiting endothelin receptor A and B with an IC50 of 15.1 1.6 M in P388/dx cells.A sulfonamide-derived, competitive and specific endothelin receptor antagonist with a relatively higher affinity to the endothelin A receptor than endothelin B receptor. Other sizes are available. Please inqury us for quote.