Learn More
Medchemexpress LLC HY-A0013A 50mg Medchemexpress, Bosentan (hydrate) CAS:157212-55-0 Purity:>98%
SDP
Catalog No. 501932891
Encompass_Preferred
Medchemexpress, HY-A0013A 50mg Bosentan (hydrate) CAS:157212-55-0 Bosentan hydrate is a competitive and dual antagonist of endothelin-1 (ET) for the ET A and ET B receptors with K i of 4.7 nM and 95 nM in human SMC, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2891 Supplier Medchemexpress LLC Supplier No. HYA0013A50MG
Medchemexpress, HY-A0013A 50mg Bosentan (hydrate) CAS:157212-55-0 Bosentan hydrate is a competitive and dual antagonist of endothelin-1 (ET) for the ET A and ET B receptors with K i of 4.7 nM and 95 nM in human SMC, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.