Learn More
Cole-Parmer IKA 2657800 Laboratory Mixer Boss Head Clamp, R 270
Supplier:  Cole-Parmer 5070397
Encompass
Get the most out of your IKA Mixer with these accessories. Made of cast aluminum. Clamping range-stand: 1" to 1 2/5"(25-36mm). Clamping range-extension arm: 1/5" to 4/5" (5-21mm).
Catalog No. NC0529887