Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Bosshead Double Clamp
SDP

Catalog No. 11392177
Change view
Click to view available options
Type:
Boss Head Double Clamp
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
11-392-177 Boss Head Double Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-177 Supplier Velp™ Supplier No. A00001301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Velp™ Bosshead Double Clamp for use with Velp™ Overhead Stirrer and OV5 Homogenizers.

  • For use with Overhead Stirrer and OV5 Homogenizers

Specifications

Color Gray
For Use With (Equipment) VELP Overhead Stirrer and OV5 homogenizers
Quantity 1 Ea.
Type Boss Head Double Clamp
Material Cast Zinc, Aluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.