Learn More
Enterprise Technology Solutions Bostitch (Stanley Bostitch) Dual Temperature Glue Sticks, 4" Sticks, 24/PK, Clear
SDP
Catalog No. 5000832426
Encompass_Preferred
Dual-temperature glue sticks are designed for use in hot-melt, low-temperature or dual-temperature glue guns. Versatile, multipurpose sticks can be used for various bonding projects from general purpose to heat-sensitive materials. Glue sticks can be used with the Stanley GR20 GlueShot glue gun and GR25-2 Dual Melt glue gun.
Catalog No. 50-008-32426 Supplier Enterprise Technology Solutions Supplier No. BOSGS20DT
Dual-temperature glue sticks are designed for use in hot-melt, low-temperature or dual-temperature glue guns. Versatile, multipurpose sticks can be used for various bonding projects from general purpose to heat-sensitive materials. Glue sticks can be used with the Stanley GR20 GlueShot glue gun and GR25-2 Dual Melt glue gun.