Learn More
Enterprise Technology Solutions Bostitch (Stanley Bostitch) Inspection Light, Ultraviolet LED, 395NM, 3/10"Wx5-1/2"L, BK
SDP
Catalog No. 5000831986
Encompass_Preferred
Ultraviolet inspection light features UV LEDs that can reveal materials that are not seen through normal lighting such as automotive leaks, pet urine and blood. Lightweight aluminum construction and handy belt clip allow easy portability. Light is powered by the two included AAA batteries and provides up to 6 hours of use. Inspection light with a 395nm output is perfect for mechanics, hunters, industrial tradesmen and more.
Catalog No. 50-008-31986 Supplier Enterprise Technology Solutions Supplier No. BOS98343
Ultraviolet inspection light features UV LEDs that can reveal materials that are not seen through normal lighting such as automotive leaks, pet urine and blood. Lightweight aluminum construction and handy belt clip allow easy portability. Light is powered by the two included AAA batteries and provides up to 6 hours of use. Inspection light with a 395nm output is perfect for mechanics, hunters, industrial tradesmen and more.