Learn More
DWK Life Sciences WHEATON™ Boston Round Bottle, Safety Coated, 250mL
SDP
Catalog No. 02927150 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
Polyethylene
Polyvinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner
02-927-150 Polyvinyl
02-927-152 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-927-150 Supplier DWK Life Sciences Supplier No. W217852
Add to Cart
Add to Cart
WHEATON™ Boston Round Bottle, Safety Coated, 250mL features a plastisol coating for added safety and is made from clear soda-lime glass conforming to USP Type III requirements. Packaged with caps attached for cleanliness.
  • Plastisol coating provides an added measure of safety
  • Made from clear soda-lime glass that conforms to USP Type III requirements
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Boston Round style available with or without caps
TRUSTED_SUSTAINABILITY
Specifications
Coating Safety Coated
Color Clear
Material Soda Lime Glass
Quantity 12/Cs.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Closure Liner Polyvinyl
Closure Material Polypropylene
Autoclavable Not Autoclavable
Capacity (English) 8 oz.
Shape Round
Disposable Reusable
Product Type Boston Round Bottle