Learn More
DWK Life Sciences WHEATON™ Boston Round Bottle, Safety Coated, Amber, 1000mL
SDP
Catalog No. 03084186 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
PTFE/Polyethylene
Polyvinyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner
03-084-186 Polyvinyl
02-927-147 PTFE/Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-084-186 Supplier DWK Life Sciences Supplier No. W217952
Add to Cart
Add to Cart
WHEATON™ Boston Round Bottle, Safety Coated, Amber, 1000mL is made from WHEATON™ 900 amber soda-lime glass conforming to USP Type III requirements. Amber glass protects light-sensitive samples, and the plastisol safety coating adds an extra measure of protection.
  • Made from Wheaton 900 amber soda-lime glass conforming to USP Type III requirements
  • Plastisol safety coating provides an added measure of protection
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Available with a choice of cap material and liner to suit application needs
TRUSTED_SUSTAINABILITY
Specifications
Coating Safety Coated
Color Amber
Material Soda Lime Glass (Type 3)
Quantity 12/Cs.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Closure Liner Polyvinyl
Closure Material Polypropylene
Autoclavable Not Autoclavable
Capacity (English) 32 oz.
Shape Round
Disposable Reusable
Product Type Boston Round Bottle