Learn More
DWK Life Sciences Wheaton™ Boston Round Bottle, Safety Coated, Amber, 250mL
Shop All DWK Life Sciences Products

Description
- Packaged in convenience packs with caps attached to maintain cleanliness
- Coated glass bottles provide an added measure of safety
- Case includes hazardous communication label and tamper evident seal if labeling and/or sealing is required
Specifications
Specifications
| Coating | Safety Coated |
| Color | Amber |
| Material | Borosilicate Glass (Type 3) |
| Quantity | 12/Cs. |
| Closure Color | White |
| Closure Included | Yes |
| Mouth | Wide |
| Product Type | Boston Round Bottle |
| Closure Liner | PTFE/Polyethylene |
| Closure Material | Polypropylene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.