Learn More
DWK Life Sciences WHEATON™ Boston Round Bottle, Safety Coated, Amber, 500mL
SDP
Catalog No. 03084185 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Liner:
PTFE/Polyethylene
Polyethylene Cone
Polyvinyl
Closure Material:
Phenolic
Polypropylene
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Liner Closure Material
03-084-185 Polyvinyl Polypropylene
02-927-143 Polyethylene Cone Phenolic
02-927-146 PTFE/Polyethylene Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-084-185 Supplier DWK Life Sciences Supplier No. W217951
Add to Cart
Add to Cart
WHEATON™ Boston Round Bottle, Safety Coated, Amber, 500mL is made from WHEATON™ 900 amber soda-lime glass conforming to USP Type III requirements. The plastisol safety coating and amber glass help protect light-sensitive samples during storage.
  • Plastisol coating provides an added measure of safety
  • Made from amber soda-lime glass conforming to USP Type III requirements
  • Amber glass protects light-sensitive samples during storage
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Available in Boston Round style
TRUSTED_SUSTAINABILITY
Specifications
Coating Safety Coated
Color Amber
Material Soda Lime Glass
Quantity 12/Cs.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Closure Liner Polyvinyl
Closure Material Polypropylene
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape Round
Disposable Reusable
Product Type Boston Round Bottle