Learn More
Uline BOSTON ROUND BOTTLES,8OZ,12/CS
Catalog No. NC2259584
Encompass
Amber Boston Round Glass Bottles - 8 ozStore and pour chemicals and medicines.Narrow mouth for precision pouring.Safe storage for hydrogen peroxide, sulfuric acid and light-sensitive chemicals.FDA compliant.Phenolic black caps included.
Catalog No. NC2259584 Supplier Uline Supplier No. S23396CS
Amber Boston Round Glass Bottles - 8 ozStore and pour chemicals and medicines.Narrow mouth for precision pouring.Safe storage for hydrogen peroxide, sulfuric acid and light-sensitive chemicals.FDA compliant.Phenolic black caps included.