Learn More
TARGETMOL CHEMICALS INC Bosutinib 200MG
SDP
Catalog No. 502880776
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Bosutinib (SKI-606) is a synthetic quinolone derivative and dual kinase inhibitor that targets both Abl (IC50 1 nM) and Src (IC50 1.2 nM) kinases. Purity 99.84%
Catalog No. 50-288-0776 Supplier TARGETMOL CHEMICALS INC Supplier No. T0152200MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Bosutinib (SKI-606) is a synthetic quinolone derivative and dual kinase inhibitor that targets both Abl (IC50 1 nM) and Src (IC50 1.2 nM) kinases. Purity 99.84%