Learn More
Selleck Chemical LLC Bosutinib S1014-200MG
SDP
Catalog No. 5000041583
Encompass_Preferred
Bosutinib is a novel dual Src/Abl inhibitor with IC50 of 1 2 nM and 1 nM in cell-free assays respectively Bosutinib also effectively decreases the activity of PI3K/AKT/mTOR MAPK/ERK and JAK/STAT3 signaling pathways by blocking the phosphorylation levels of p-ERK p-S6 and p-STAT3 Bosutinib promotes autophagy
Catalog No. 50-000-41583 Supplier Selleck Chemical LLC Supplier No. S1014200MG
Bosutinib is a novel dual Src/Abl inhibitor with IC50 of 1 2 nM and 1 nM in cell-free assays respectively Bosutinib also effectively decreases the activity of PI3K/AKT/mTOR MAPK/ERK and JAK/STAT3 signaling pathways by blocking the phosphorylation levels of p-ERK p-S6 and p-STAT3 Bosutinib promotes autophagy