Learn More
Medchemexpress LLC Bosutinib hydrate | 918639-08-4 | MFCD29920031 | 100.0% | 548.46 g/mol | C26H31Cl2N5O4 | 200 MG
SDP
Catalog No. 5000245673
Encompass_Preferred
Bosutinib hydrate is a small-molecule dual Src/Abl tyrosine kinase inhibitor supplied as a white to off-white solid for research use. It exhibits low-nanomolar potency against Src and Abl kinases and is intended for biochemical and cell-based studies; supporting documents such as SDS and COA are provided for analytical verification and safe handling.
  • Dual Src and Abl kinase inhibition at low-nanomolar concentrations.
  • High reported purity suitable for research applications.
  • Available in multiple packaging sizes, including 200 MG.
  • White to off-white solid appearance for easy handling.
  • Storage and stability guidance provided for solid and solution forms.
  • COA and SDS available to support quality and safety use.
Catalog No. 50-002-45673 Supplier Medchemexpress LLC Supplier No. HY10158A200MG
Bosutinib hydrate is a small-molecule dual Src/Abl tyrosine kinase inhibitor supplied as a white to off-white solid for research use. It exhibits low-nanomolar potency against Src and Abl kinases and is intended for biochemical and cell-based studies; supporting documents such as SDS and COA are provided for analytical verification and safe handling.
  • Dual Src and Abl kinase inhibition at low-nanomolar concentrations.
  • High reported purity suitable for research applications.
  • Available in multiple packaging sizes, including 200 MG.
  • White to off-white solid appearance for easy handling.
  • Storage and stability guidance provided for solid and solution forms.
  • COA and SDS available to support quality and safety use.