Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bosutinib isomer | 1391063-17-4 | MFCD22418831 | 99.6% | 530.45 g·mol⁻¹ | C26H29Cl2N5O3 | 25 MG
SDP

Catalog No. 5000246318
Encompass_Preferred

Bosutinib isomer is a small-molecule kinase inhibitor that binds Wee1 and Wee2 with high affinity and is intended for biochemical and cellular research applications. It is supplied as a solid and is typically dissolved in DMSO for preparation of stock solutions.

  • High binding affinity for Wee1 (Kd 43.7 ± 10.0 nM) and Wee2 (Kd 4.7 ± 2.3 nM).
  • Purity 99.62%.
  • Chemical formula C26H29Cl2N5O3; molecular weight 530.45 g·mol⁻¹.
  • Provided in milligram-scale quantities suitable for assay development and screening.
  • Soluble in DMSO for assay preparation.
  • For research use only; not for human or animal therapeutic use.

Catalog No. 50-002-46318 Supplier Medchemexpress LLC Supplier No. HY10302925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bosutinib isomer is a small-molecule kinase inhibitor that binds Wee1 and Wee2 with high affinity and is intended for biochemical and cellular research applications. It is supplied as a solid and is typically dissolved in DMSO for preparation of stock solutions.

  • High binding affinity for Wee1 (Kd 43.7 ± 10.0 nM) and Wee2 (Kd 4.7 ± 2.3 nM).
  • Purity 99.62%.
  • Chemical formula C26H29Cl2N5O3; molecular weight 530.45 g·mol⁻¹.
  • Provided in milligram-scale quantities suitable for assay development and screening.
  • Soluble in DMSO for assay preparation.
  • For research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.