Learn More
Cayman Chemical β-Boswellic Acid, Purity: ≥95%, CAS: 631-69-6, 10 mg
SDP
Catalog No. 502951757
Encompass_Preferred
A pentacyclic triterpene with diverse bioactivities; inhibits 5-LO (IC50 = ~5 µM); inhibits cell proliferation in HL-60 cells (IC50s = 3.7, 7.1, and 6.3 µM for DNA, RNA, and protein synthesis, respectively); increases nitric oxide and phosphorylation of eNOS in HUVECs deprived of oxygen and glucose; increases calcium mobilization in platelets (≥3 µM); induces platelet aggregation (EC50 = 8 µM)
Catalog No. 50-295-1757 Supplier Cayman Chemical Supplier No. 1169310 MG
A pentacyclic triterpene with diverse bioactivities; inhibits 5-LO (IC50 = ~5 µM); inhibits cell proliferation in HL-60 cells (IC50s = 3.7, 7.1, and 6.3 µM for DNA, RNA, and protein synthesis, respectively); increases nitric oxide and phosphorylation of eNOS in HUVECs deprived of oxygen and glucose; increases calcium mobilization in platelets (≥3 µM); induces platelet aggregation (EC50 = 8 µM)