Learn More
MSC Bioesque Solutions Disinfectant, Broad-Spectrum, EPA List N, 5 gal
Catalog No. 502868457
Encompass
This botanical disinfectant solution sanitizes, disinfects, cleans, and deodorizes. It is on EPA's List N to kill SARS-CoV-2 (COVID-19) and is tested and approved for use with electrostatic sprayers with a particle size ≥40 microns in diameter.
  • Kills SARS-CoV-2 virus (COVID-19) in 1 minute on hard non-porous surfaces
  • Kills 99.9% of bacteria, viruses, fungi, and molds
  • EPA registered broad-spectrum disinfectant (bactericidal, virucidal, tuberculocidal, fungicidal)
  • No rinse required, even on food contact surfaces
  • Sanitizes soft surfaces in 2 minutes
  • Eliminates 99.9% of most allergens
  • Non-abrasive, non-corrosive, non-flammable formula
  • Free from harsh chemicals, chlorine, and phosphates; no PPE needed
  • Ready-to-use with pleasant lemongrass grapefruit scent
  • Botanically derived active ingredient, effective for restoration and remediation
Catalog No. 50-286-8457 Supplier MSC Supplier No. 19420496
This botanical disinfectant solution sanitizes, disinfects, cleans, and deodorizes. It is on EPA's List N to kill SARS-CoV-2 (COVID-19) and is tested and approved for use with electrostatic sprayers with a particle size ≥40 microns in diameter.
  • Kills SARS-CoV-2 virus (COVID-19) in 1 minute on hard non-porous surfaces
  • Kills 99.9% of bacteria, viruses, fungi, and molds
  • EPA registered broad-spectrum disinfectant (bactericidal, virucidal, tuberculocidal, fungicidal)
  • No rinse required, even on food contact surfaces
  • Sanitizes soft surfaces in 2 minutes
  • Eliminates 99.9% of most allergens
  • Non-abrasive, non-corrosive, non-flammable formula
  • Free from harsh chemicals, chlorine, and phosphates; no PPE needed
  • Ready-to-use with pleasant lemongrass grapefruit scent
  • Botanically derived active ingredient, effective for restoration and remediation