Learn More
Botany Survey Set
SCI_ED
Catalog No. S10013A
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
S10013A 25 Pk.
1 options
Catalog No. S10013A Supplier Triarch Inc Supplier No. S10013A
Only null left
Add to Cart
Add to Cart
Includes
  • Bacteria, three shapes, individual smears
  • Stemonitis (slime mold), w.m. of sporangia
  • Mold, three types, w.m.
  • Mushrooms, c.s.
  • Yeast, budding, w.m.
  • Nostoc, w.m.
  • Diatoms, mixed, w.m.
  • Spirogyra, vegetating and conjugating, w.m.
  • Euglena, w.m.
  • Fucus, c.s. of monoecious conceptacle
  • Polysiphonia, w.m. with tetraspores
  • Fern spores, germinating, w.m.
  • Fern rhizome, c.s.
  • Pine leaf, c.s.
  • Clover nodule, c.s.
  • Basswood stem, c.s.
  • Monocot and dicot roots, c.s.
  • Monocot and dicot stems, c.s.
  • Monocot and dicot leaves, c.s.
  • Onion root tip, l.s., mitosis
  • Root hairs, w.m. of grass root tip
  • Corn stem, c.s.
  • Elodea leaf, w.m.
  • Leaf types, c.s. of hydrophyte, mesophyte and xerophyte types
  • Pollen types, w.m. of five kinds
This 25-slide set ranges from bacteria through higher plants.
Specifications
Type Botany Survey
Quantity 25 Pk.