Learn More
Cayman Chemical Botn-HPDP 5mg
SDP
Catalog No. 502970448
Encompass_Preferred
A sulfhydryl-reactive biotinylation reagent that forms a reversible disulfide linkage, used to label protein cysteines and other substrates that contain sulfhydryl groups; EAsily linked with target substrates at pH 6.5 to 7.5 in buffers such as PBS; disulfide linkage can be cleaved by reducing agents
Catalog No. 50-297-0448 Supplier Cayman Chemical Supplier No. 164595 MG
A sulfhydryl-reactive biotinylation reagent that forms a reversible disulfide linkage, used to label protein cysteines and other substrates that contain sulfhydryl groups; EAsily linked with target substrates at pH 6.5 to 7.5 in buffers such as PBS; disulfide linkage can be cleaved by reducing agents