Learn More
Thomas Scientific Centrifuge Bottle, PP, 500mL, Seal Cap
SDP
Catalog No. 501458323
Encompass
Foxx Life Sciences Autofil® Centrifuge Bottles are designed specifically to assist Life Science researchers in processing samples for cell suspensions, cell lysates, tissue homogenates, and organ homogenates. Autofil® Centrifuge Bottles are available in 250ml or 500ml and fit a wide range of rotors. Material options include polycarbonate or polypropylene with your choice of a traditional screw cap or a leak-proof sealing cap with silicone O-ring. Our centrifuge bottles feature thick walls that hold up to reuse along with wide mouths that simplify filling and pouring liquids. Each Autofil® Centrifuge Bottle also features large, clear-molded graduation markings and a frosted area for convenient labeling.
Catalog No. 50-145-8323 Supplier Thomas Scientific Supplier No. 1171Z70
Foxx Life Sciences Autofil® Centrifuge Bottles are designed specifically to assist Life Science researchers in processing samples for cell suspensions, cell lysates, tissue homogenates, and organ homogenates. Autofil® Centrifuge Bottles are available in 250ml or 500ml and fit a wide range of rotors. Material options include polycarbonate or polypropylene with your choice of a traditional screw cap or a leak-proof sealing cap with silicone O-ring. Our centrifuge bottles feature thick walls that hold up to reuse along with wide mouths that simplify filling and pouring liquids. Each Autofil® Centrifuge Bottle also features large, clear-molded graduation markings and a frosted area for convenient labeling.