Learn More
Thermo Scientific BOTTLE 2 L PLASTIC
Catalog No. NC9881156 Shop All Thermo Scientific Products
Encompass
2L Eluent Bottle, Plastic, Non-twist stopper with separate retaining ring to prevent tubing from tangling; custom-molded polyethylene end-line filters with 5�m pores; Graduations marked on containers in 100mL increments
Catalog No. NC9881156 Supplier Thermo Scientific Supplier No. 062510
2L Eluent Bottle, Plastic, Non-twist stopper with separate retaining ring to prevent tubing from tangling; custom-molded polyethylene end-line filters with 5�m pores; Graduations marked on containers in 100mL increments