Learn More
Ace Glass, Inc. Bottle, pressure, 325mL, 60psi, complete with #25 back-seal plug
SDP
Catalog No. 50979813
Encompass_Preferred
325mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Includes #25 back-seal plug.
Catalog No. 50-979-813 Supplier Ace Glass, Inc. Supplier No. 8648248
325mL pressure bottle with #25 Ace-Thred, rated for 60psig at room temperature. Includes #25 back-seal plug.