Learn More
Ace Glass, Inc. Bottle, pressure, 325mL, 60psi, complete with #36 front-seal plug
SDP
Catalog No. 50979814
Encompass_Preferred
325mL pressure bottle with #36 Ace-Thred, rated for 60psig at room temperature. Includes #36 front-seal plug.
Catalog No. 50-979-814 Supplier Ace Glass, Inc. Supplier No. 8648308
325mL pressure bottle with #36 Ace-Thred, rated for 60psig at room temperature. Includes #36 front-seal plug.