Learn More
Ace Glass, Inc. Bottle, pressure, 325mL, 60psi, complete with #36 back-seal plug
SDP
Supplier:  Ace Glass, Inc. 8648309
Encompass_Preferred
325mL pressure bottle with #36 Ace-Thred, rated for 60psig at room temperature. Includes #36 back-seal plug.
Catalog No. 50-976-224