Learn More
Chemglass Life Sciences BelloCell -500P Cell Culture Bottle
SDP
Catalog No. 501532097
Encompass
High Cell Density (up to 5 x10(9) Cells/Bottle).BelloCell-500P is an alternative choice with automatic medium recirculation that does not require manual culture media exchange. It reduces chances of contamination, and maximizes stability by supplying sufficient culture medium. With the aid of the BelloFeeder pump, the inlet and outlet ports allowmedia to flow from and to the associated medium reservoir that provides sufficient nutrient and stable pH during culture.
Catalog No. 50-153-2097 Supplier Chemglass Life Sciences Supplier No. CLS130211
High Cell Density (up to 5 x10(9) Cells/Bottle).BelloCell-500P is an alternative choice with automatic medium recirculation that does not require manual culture media exchange. It reduces chances of contamination, and maximizes stability by supplying sufficient culture medium. With the aid of the BelloFeeder pump, the inlet and outlet ports allowmedia to flow from and to the associated medium reservoir that provides sufficient nutrient and stable pH during culture.