Learn More
Chemglass Life Sciences BelloCell -500 Cell Culture Bottle
SDP
Catalog No. 501532096
Encompass
BelloCell -500 Cell Culture Bottle. BelloCell-500 was the first bottle available to the market, introducing theconcept of tidal flow for high-density cell culture. The simple design combines multiple functions of a mixer, oxygenator, and a large growing surface to achieve a high density of cells. The BelloCell-500 provides unlimited oxygen during culture with relatively gentle movement of the culture medium. Porous carriers are packed in the bottle that provides 15,600 cm surface area for cell growth. Its space and labor savings are the most notable features. One BelloCell-500 is equivalent to dozens of roller bottles, and occupies only 1/20th the space and labor. It is the most user friendly tool for laboratory researchers.
Catalog No. 50-153-2096 Supplier Chemglass Life Sciences Supplier No. CLS130010
BelloCell -500 Cell Culture Bottle. BelloCell-500 was the first bottle available to the market, introducing theconcept of tidal flow for high-density cell culture. The simple design combines multiple functions of a mixer, oxygenator, and a large growing surface to achieve a high density of cells. The BelloCell-500 provides unlimited oxygen during culture with relatively gentle movement of the culture medium. Porous carriers are packed in the bottle that provides 15,600 cm surface area for cell growth. Its space and labor savings are the most notable features. One BelloCell-500 is equivalent to dozens of roller bottles, and occupies only 1/20th the space and labor. It is the most user friendly tool for laboratory researchers.