Learn More
Chemglass Life Sciences Bottle, 500mL, BelloCell-500A
SDP
Catalog No. 501532098
Encompass
High Cell Density (3~5x10(9) Cells/Bottle).BelloCell-500A is designed to remove the lid on the matrix basket allowing for easier collection of the carriers. Inverting the bottle, carriers fall into the cap and provide special functions such as cell seeding, virus infection, transfection, transduction, and cell harvest (solid screw cap is provided for this purpose). This is particularly ideal for extracting cellular protein or viruses directly from the carriers. MAJOR NOTABLE FEATURES: To increase cell adherence efficiency for certain cell lines that are sensitive to trypsinization by inverting the bottles, especially HEK293. To enhance virus infection/transfection/transduction efficiency with high virus or plasmid concentration by inverting the bottles. To save trypsin solution and increase cell harvest efficiency under sterile condition.
Catalog No. 50-153-2098 Supplier Chemglass Life Sciences Supplier No. CLS130612
High Cell Density (3~5x10(9) Cells/Bottle).BelloCell-500A is designed to remove the lid on the matrix basket allowing for easier collection of the carriers. Inverting the bottle, carriers fall into the cap and provide special functions such as cell seeding, virus infection, transfection, transduction, and cell harvest (solid screw cap is provided for this purpose). This is particularly ideal for extracting cellular protein or viruses directly from the carriers. MAJOR NOTABLE FEATURES: To increase cell adherence efficiency for certain cell lines that are sensitive to trypsinization by inverting the bottles, especially HEK293. To enhance virus infection/transfection/transduction efficiency with high virus or plasmid concentration by inverting the bottles. To save trypsin solution and increase cell harvest efficiency under sterile condition.