Learn More
Ace Glass, Inc. Bottle, pressure, 950mL, 60psi, complete with #25 back-seal plug
SDP
Catalog No. 50976222
Encompass_Preferred
950mL pressure bottle with #25 Ace-Thred, rated for 60psig at as much as 120C. Includes #25 back-seal plug.
Catalog No. 50-976-222 Supplier Ace Glass, Inc. Supplier No. 8648252
950mL pressure bottle with #25 Ace-Thred, rated for 60psig at as much as 120C. Includes #25 back-seal plug.