Learn More
Medi Dose Inc Eps Inc BOTTLE ADAPTER 17-20MM OPENING
SDP
Catalog No. NC0866135
Encompass
Turns any bottle into a ready-to-dispense medication reservoir! Min opening -17 mm Maximum opening 20 mm. 100 Adapters
Catalog No. NC0866135 Supplier Medi Dose Inc Eps Inc Supplier No. PBA24
Turns any bottle into a ready-to-dispense medication reservoir! Min opening -17 mm Maximum opening 20 mm. 100 Adapters