Learn More
Chemglass Life Sciences 500mL Bottle, 53-400 GPI Thread, Amber Glass
SDP
Catalog No. CG82004
Encompass_Preferred
500mL Bottle, 53-400 GPI Thread, Amber Glass. These round, wide mouth amber glass bottles are used for storage of light sensitive chemicals. Has a white polypropylene cap with a polyethylene faced foam liner. Sold with cap attached to the bottle in case quantities only.
Catalog No. CG82004 Supplier Chemglass Life Sciences Supplier No. CG82004
500mL Bottle, 53-400 GPI Thread, Amber Glass. These round, wide mouth amber glass bottles are used for storage of light sensitive chemicals. Has a white polypropylene cap with a polyethylene faced foam liner. Sold with cap attached to the bottle in case quantities only.