Learn More
Chemglass Life Sciences Bottle, Anaerobic, 2000mL, 2-Neck
SDP
Catalog No. 501532361
Encompass_Preferred
Bottle, Anaerobic, 2000mL, 2-Neck. Bottle is convenient for growing cultures in that same manner as the CLS-4209-01 anaerobic culture tubes.
Catalog No. 50-153-2361 Supplier Chemglass Life Sciences Supplier No. CLS421501
Bottle, Anaerobic, 2000mL, 2-Neck. Bottle is convenient for growing cultures in that same manner as the CLS-4209-01 anaerobic culture tubes.