Learn More
Chemglass Life Sciences Bottle, Anaerobic, 500mL, Single Neck
SDP
Catalog No. 501532362
Encompass_Preferred
Bottle, Anaerobic, 500mL, Single Neck. Bottle is convenient for growing cultures in that same manner as the CLS-4209-01 anaerobic culture tubes.
Catalog No. 50-153-2362 Supplier Chemglass Life Sciences Supplier No. CLS421503
Bottle, Anaerobic, 500mL, Single Neck. Bottle is convenient for growing cultures in that same manner as the CLS-4209-01 anaerobic culture tubes.