Learn More
Chemglass Life Sciences Bottle, Anaerobic, Media, 50mL, Aluminum Seal
SDP
Catalog No. 501532364
Encompass_Preferred
Bottle, Anaerobic, Media, 50mL, Aluminum Seal. Anaerobic media bottles are designed for maintaining anaerobic conditions, useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development. The bottle is manufactured from Duran borosilicate glass, autoclavable and each has a graduation scale indicating the approximate volumes with a writing area for identification. Supplied with a rubber stopper and aluminum seal.
Catalog No. 50-153-2364 Supplier Chemglass Life Sciences Supplier No. CLS421701
Bottle, Anaerobic, Media, 50mL, Aluminum Seal. Anaerobic media bottles are designed for maintaining anaerobic conditions, useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development. The bottle is manufactured from Duran borosilicate glass, autoclavable and each has a graduation scale indicating the approximate volumes with a writing area for identification. Supplied with a rubber stopper and aluminum seal.