Learn More
Beckman Coulter 1 L (1000 mL) Polypropylene Bottle Assembly, 2/pk
Catalog No. NC2001061
Encompass
1L (1000mL) Polypropylene Bottle for use in High Performance and High Capacity Rotors JLA-9.1000 and JLA-8.1000.
Catalog No. NC2001061 Supplier Beckman Coulter Supplier No. C31597
1L (1000mL) Polypropylene Bottle for use in High Performance and High Capacity Rotors JLA-9.1000 and JLA-8.1000.