Learn More
Chemglass Life Sciences Bottle Assembly, 125mL, 2-Port, Single-Use AdvantaFlex Tubing, with Male Luer, Bioprocess, Single Use
SDP
Catalog No. 502733337
Encompass
Strain relief molded into stopper at tubing exit areas to prevent kinking.Manufactured, assembled and packaged in Class 7 cleanroom.USP Class VI approved materials.Animal Derived Component Free (ADCF).Gamma irradiated at 25kGY-40kGY.Oursingle-use bottle assemblies are available in a wide range of standard sizes, from 125mL to 2L.Each bottle assembly includes a PETG bottle, a 2-port closure assembly that includes a 30mm 0. 22uM vent filter, a 24-inch length of tubing with a plugged end, a 1/4 pinch clamp, a V-notched dip tube that extends to the bottom of the bottle and a polypropylene cap.The assembly is made from pharmaceutical grade thermoplastic elastomer (TPE) resin.The entire system is designed to ensure an unobstructed fluid path.Each unit is double poly-bagged and Gamma Irradiated at 25kGY-35kGY.
Catalog No. 50-273-3337 Supplier Chemglass Life Sciences Supplier No. CLS11680125MLC
Strain relief molded into stopper at tubing exit areas to prevent kinking.Manufactured, assembled and packaged in Class 7 cleanroom.USP Class VI approved materials.Animal Derived Component Free (ADCF).Gamma irradiated at 25kGY-40kGY.Oursingle-use bottle assemblies are available in a wide range of standard sizes, from 125mL to 2L.Each bottle assembly includes a PETG bottle, a 2-port closure assembly that includes a 30mm 0. 22uM vent filter, a 24-inch length of tubing with a plugged end, a 1/4 pinch clamp, a V-notched dip tube that extends to the bottom of the bottle and a polypropylene cap.The assembly is made from pharmaceutical grade thermoplastic elastomer (TPE) resin.The entire system is designed to ensure an unobstructed fluid path.Each unit is double poly-bagged and Gamma Irradiated at 25kGY-35kGY.