Learn More
Chemglass Life Sciences Chemglass Bottle Assembly, 500 mL, 2-Port, Single-Use (24 in tubing, 30 mm 0.22 μm vent filter) - Pack of 5
SDP
Catalog No. NC3710845
Encompass_Preferred
The Chemglass 500 mL bottle assembly is a single-use PETG media transfer and feed bottle configured with a two-port closure, vent filter, and welded tubing for sterile fluid handling. It is assembled and packaged for immediate use in laboratory and bioprocess workflows.
  • 500 mL capacity
  • PETG bottle material
  • Two-port closure assembly
  • 30 mm 0.22 μm vent filter
  • 24 in of 1/4 in OD weldable tubing with plugged end
  • 1/4 in pinch clamp included
  • V-notched dip tube extends to bottom of bottle
  • Polypropylene cap included
  • Pack of 5, double poly-bagged and gamma irradiated (25-35 kGy)
  • Manufactured and packaged in a Class 7 cleanroom
Catalog No. NC3710845 Supplier Chemglass Life Sciences Supplier No. CLS11680500CFXMS50117
The Chemglass 500 mL bottle assembly is a single-use PETG media transfer and feed bottle configured with a two-port closure, vent filter, and welded tubing for sterile fluid handling. It is assembled and packaged for immediate use in laboratory and bioprocess workflows.
  • 500 mL capacity
  • PETG bottle material
  • Two-port closure assembly
  • 30 mm 0.22 μm vent filter
  • 24 in of 1/4 in OD weldable tubing with plugged end
  • 1/4 in pinch clamp included
  • V-notched dip tube extends to bottom of bottle
  • Polypropylene cap included
  • Pack of 5, double poly-bagged and gamma irradiated (25-35 kGy)
  • Manufactured and packaged in a Class 7 cleanroom