Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Bottle Assy J-Lite 1000ml PC (set of 2)

Catalog No. 501631980
Encompass_Preferred

Bottle Assembly, J-Lite, Polycarbonate, 1000mL, Quantity: Set of 2

Catalog No. 50-163-1980 Supplier Beckman Coulter Supplier No. A98812

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Bottle Assembly, J-Lite, Polycarbonate, 1000mL, Quantity: Set of 2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.