Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bottle Biology
SCI_ED

Catalog No. S83132
Encompass
Encompass_Preferred
Change view
Click to view available options
Format:
Book
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S83132 Book
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S83132 Supplier KENDALL HUNT PUBLISHING COMPANY Supplier No. 9780757500947
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Explore science and the environment using recyclable materials!

Model a rainforest, create spider habitat, observe the slime mold lifecycle, and explore an ecosystem. Pursue these and other scientific investigations with over 20 bottle constructions, including the Ecocolumn, Predator-Prey Column, Niche Kit, and TerrAqua Column. Spiral bound. Soft cover. 127 pages. 1993.

Specifications

Format Book
Product Type Bottle Biology
Includes Ecocolumn, predator-prey column, niche kit and TerrAqua column
For Use With (Application) Explores science and the environment using recyclable materials
Subject Biology
Author Wisconsin Fast Plants Program
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.