Learn More
Andwin Scientific BOTTLE BRUSH
SDP
Catalog No. NC9451201
Encompass_Preferred
The Babcock milk test Bottle Brush is specially designed with extra long natural bristles that can perfectly and conveniently fit Babcock milk test bottles that have narrow necks. The short dense bristles along the stem allows the users to efficiently scrub and clean the bottles. This product is sold in a pack of 5.
Catalog No. NC9451201 Supplier Andwin Scientific Supplier No. 17068009
The Babcock milk test Bottle Brush is specially designed with extra long natural bristles that can perfectly and conveniently fit Babcock milk test bottles that have narrow necks. The short dense bristles along the stem allows the users to efficiently scrub and clean the bottles. This product is sold in a pack of 5.