Learn More
Cole-Parmer Small Bottle/cylinder Brush, 11-1/2", 12/pk
Supplier:  Cole-Parmer 8455015
Encompass_Preferred
Ideal for cleaning bottles, cylinders, and large test tubes. Full radial end tip promotes effective cleaning and reduces scratching of glass and plasticware. Wire handle has hanging hook.
Catalog No. NC0713494