Learn More
Peeke Scientific GL45 Cap, with Integrated Air Inlet Valve
Catalog No. NC1453616
Encompass_Preferred
BOTTLE CAP, GL45, (4) 1/4-28 ports, PTFE Center with green PP collar, EPDM air inlet valve.
Catalog No. NC1453616 Supplier Peeke Scientific Supplier No. GL45428E
BOTTLE CAP, GL45, (4) 1/4-28 ports, PTFE Center with green PP collar, EPDM air inlet valve.