Learn More
Beckman Coulter Beckman Coulter BOTTLE/CAP, PC 29X104 50ML (PKG-25
Catalog No. 501631976
Encompass_Preferred
50 mL Polycarbonate Bottle/Cap. Dimensions: 29X104mm, Package of 25
Catalog No. 50-163-1976 Supplier Beckman Coulter Supplier No. 357002
50 mL Polycarbonate Bottle/Cap. Dimensions: 29X104mm, Package of 25