Learn More
Waters Corp Waters Bottle Cap, Polyethylene, 1.25 in. Neck, 4 L, 4/pk
Catalog No. 50818888 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polyethylene bottle cap is designed to prevent the free exchange of atmospheric gases with helium. It will not sustain an internal pressure build-up and features three holes to accommodate various lines for HPLC systems.
  • Polyethylene cap prevents free exchange of atmospheric gases with helium.
  • Will not sustain an internal pressure build-up.
  • Three-holed cap.
  • Accommodates a helium inlet line, helium exhaust line, and a solvent inlet line.
Catalog No. 50-818-888 Supplier Waters Corp Supplier No. WAT062341
This polyethylene bottle cap is designed to prevent the free exchange of atmospheric gases with helium. It will not sustain an internal pressure build-up and features three holes to accommodate various lines for HPLC systems.
  • Polyethylene cap prevents free exchange of atmospheric gases with helium.
  • Will not sustain an internal pressure build-up.
  • Three-holed cap.
  • Accommodates a helium inlet line, helium exhaust line, and a solvent inlet line.