Learn More
Hach Company Carboy, with Spigot, 4-L Capacity
Supplier:  Hach Company 1486817
Encompass
Low-density polyethylene with a leak-proof polypropylene screw closure. Molded in spigot provides strength and leak-proof operation. 10 L and20 L sizes have wide shoulder handles.
Catalog No. 50-199-5141