Learn More
BURKLE INC Bottle carrier for 6 bottles up to max. Ø 3.74 in. (95 mm)
Catalog No. 5000025604
Encompass_Preferred
The Burkle bottle carrier is the ideal solution for the safe and efficient transportation of bottles in laboratories and other demanding environments.
It is made of chemical-resistant and food-safe polypropylene (PP) and offers space for up to six bottles with a maximum diameter of 95 mm (3.74 in.).
Thanks to its compact design, it is space-saving to store and easy to transport. The robust construction ensures durability and reliability.
The bottle carrier minimizes the risk of breakage or spillage, enables hygienic handling of containers and saves valuable time by transporting several bottles at the same time.
Catalog No. 50-000-25604 Supplier BURKLE INC Supplier No. 03610006
The Burkle bottle carrier is the ideal solution for the safe and efficient transportation of bottles in laboratories and other demanding environments.
It is made of chemical-resistant and food-safe polypropylene (PP) and offers space for up to six bottles with a maximum diameter of 95 mm (3.74 in.).
Thanks to its compact design, it is space-saving to store and easy to transport. The robust construction ensures durability and reliability.
The bottle carrier minimizes the risk of breakage or spillage, enables hygienic handling of containers and saves valuable time by transporting several bottles at the same time.