Learn More
Thomas Scientific Bottle Carrier, Black, Small
SDP
Catalog No. NC1534708
Encompass_Preferred
Resists most aqueous solutions, acids and bases Steam sterilizable Can be used from -45Ý C to 110Ý C Centering ring holds bottle from wall Large carrier accommodates 2.5 L and 4 L bottles. Small carrier accommodates 500 mL and 1,000 mL bottles. Made from a unique, high performance, thermoplastic elastomer. Handle is molded as part of the carrier to assure uniform appearance and quality. Carries liquids and ice without sweating. Resists bacteria and fungi growth. Available in three colors.
Catalog No. NC1534708 Supplier Thomas Scientific Supplier No. 1784M26
Resists most aqueous solutions, acids and bases Steam sterilizable Can be used from -45Ý C to 110Ý C Centering ring holds bottle from wall Large carrier accommodates 2.5 L and 4 L bottles. Small carrier accommodates 500 mL and 1,000 mL bottles. Made from a unique, high performance, thermoplastic elastomer. Handle is molded as part of the carrier to assure uniform appearance and quality. Carries liquids and ice without sweating. Resists bacteria and fungi growth. Available in three colors.